Product Details

SNP ID
rs7112680
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:69294610 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGGTCTTTCTTATGTTTTAAGGA[A/G]AGCCCTGTGGCTGCTGGGTGGGGGA
Phenotype
MIM: 605625
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYEOV PubMed Links

Gene Details

Gene
MYEOV
Gene Name
myeloma overexpressed
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293291.1 Intron NP_001280220.1
NM_001293294.1 Intron NP_001280223.1
NM_001293296.1 Intron NP_001280225.1
NM_001300923.1 Intron NP_001287852.1
NM_001300924.1 Intron NP_001287853.1
NM_138768.3 Intron NP_620123.2

View Full Product Details