Product Details

SNP ID
rs61745407
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124542834 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGACTATTATGGTATAGAACAGG[A/G]AGGACACTTTCCCTTGCTCGAGGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR8B12 PubMed Links

Gene Details

Gene
OR8B12
Gene Name
olfactory receptor family 8 subfamily B member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005195.1 821 Missense Mutation TCC,TTC S274F NP_001005195.1

View Full Product Details