Product Details

SNP ID
rs568493740
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4352876 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGCTGAGGGTCCCACCGCTGTC[C/T]CTGACCCCTAACCCCTGCAGAGCCC
Phenotype
MIM: 601768
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MPND PubMed Links

Gene Details

Gene
MPND
Gene Name
MPN domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159846.2 Intron NP_001153318.1
NM_001300862.1 Intron NP_001287791.1
NM_032868.5 Intron NP_116257.2
XM_006722926.2 Intron XP_006722989.1
Gene
SH3GL1
Gene Name
SH3 domain containing GRB2 like 1, endophilin A2
There are no transcripts associated with this gene.

View Full Product Details