Product Details

SNP ID
rs59725214
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:167809759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGAATGTTTACCCTGCCTGCTAC[A/G]ATTTTTTCCCATGATGGGAGACTCA
Phenotype
MIM: 605205
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADCY10 PubMed Links

Gene Details

Gene
ADCY10
Gene Name
adenylate cyclase 10, soluble
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167749.2 6229 Silent Mutation ATA,ATG I1431M NP_001161221.1
NM_001297772.1 6229 Silent Mutation ATA,ATG I1492M NP_001284701.1
NM_018417.5 6229 Silent Mutation ATA,ATG I1584M NP_060887.2
XM_005245330.4 6229 Intron XP_005245387.1
XM_006711449.3 6229 Silent Mutation ATA,ATG I1521M XP_006711512.1
XM_011509760.2 6229 Missense Mutation ATA,ATG I1584M XP_011508062.1
XM_011509762.2 6229 Missense Mutation ATA,ATG I1536M XP_011508064.1
XM_011509763.2 6229 Missense Mutation ATA,ATG I1530M XP_011508065.1
XM_011509766.2 6229 Intron XP_011508068.1
XM_017001778.1 6229 Intron XP_016857267.1

View Full Product Details