Product Details

SNP ID
rs41363545
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18279901 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGAAAGAACCGAAGGGGGGAGCGA[C/G]ATCGAGGAAAGGGGCGGAGAATTTT
Phenotype
MIM: 165162
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
JUND PubMed Links

Gene Details

Gene
JUND
Gene Name
JunD proto-oncogene, AP-1 transcription factor subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286968.1 1756 UTR 3 NP_001273897.1
NM_005354.5 1756 UTR 3 NP_005345.3
Gene
KIAA1683
Gene Name
KIAA1683
There are no transcripts associated with this gene.

Gene
MIR3188
Gene Name
microRNA 3188
There are no transcripts associated with this gene.

View Full Product Details