Product Details

SNP ID
rs186256592
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:46837057 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTTCACGCCACACTCTAGGAAGT[C/G]CCTGAGCTAGTGAGCTGGCCAACGA
Phenotype
MIM: 300757
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RP2 PubMed Links

Gene Details

Gene
RP2
Gene Name
retinitis pigmentosa 2 (X-linked recessive)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006915.2 146 UTR 5 NP_008846.2

View Full Product Details