Product Details

SNP ID
rs17775790
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:102828366 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAAACTGGTAGGTTTAAATCTGGA[C/T]ACATTATCTAAATCTCCCATTATCC
Phenotype
MIM: 607909
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AZIN1 PubMed Links

Gene Details

Gene
AZIN1
Gene Name
antizyme inhibitor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301668.1 1927 UTR 3 NP_001288597.1
NM_015878.5 1927 UTR 3 NP_056962.2
NM_148174.3 1927 UTR 3 NP_680479.1
XM_005250969.1 1927 UTR 3 XP_005251026.1
XM_011517126.1 1927 UTR 3 XP_011515428.1
XM_011517127.1 1927 UTR 3 XP_011515429.1
XM_011517128.2 1927 UTR 3 XP_011515430.1
XM_017013563.1 1927 UTR 3 XP_016869052.1
XM_017013564.1 1927 UTR 3 XP_016869053.1
XM_017013565.1 1927 UTR 3 XP_016869054.1
XM_017013566.1 1927 UTR 3 XP_016869055.1
Gene
LOC107986873
Gene Name
forkhead box protein L2-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017014121.1 1927 Intron XP_016869610.1

View Full Product Details