Product Details

SNP ID
rs3827440
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:79171491 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTCCTCAGAACCAGTGATGATACC[T/C]CTGGCAATAGGACCAAATGCTTTGT
Phenotype
MIM: 300903
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
GPR174 PubMed Links

Gene Details

Gene
GPR174
Gene Name
G protein-coupled receptor 174
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032553.1 520 Missense Mutation CCT,TCT P162S NP_115942.1

View Full Product Details