Product Details

SNP ID
rs7970908
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120098979 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAGCGCAGCCCTGAGGGGCGCAGC[C/T]GGCTGCCTCTCTCCCACCCAACCCC
Phenotype
MIM: 617002 MIM: 604199
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BICDL1 PubMed Links

Gene Details

Gene
BICDL1
Gene Name
BICD family like cargo adaptor 1
There are no transcripts associated with this gene.

Gene
RAB35
Gene Name
RAB35, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167606.1 Intron NP_001161078.1
NM_006861.6 Intron NP_006852.1
XM_011537779.1 Intron XP_011536081.1
XM_017018723.1 Intron XP_016874212.1

View Full Product Details