Product Details

SNP ID
rs113443076
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24132344 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCCGACTGGTAGTAGGGGCAGCC[A/G]TGTGGCGGGGAGCCGGCCGGGCCTT
Phenotype
MIM: 613317 MIM: 612028 MIM: 600654
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCAF11 PubMed Links

Gene Details

Gene
DCAF11
Gene Name
DDB1 and CUL4 associated factor 11
There are no transcripts associated with this gene.

Gene
EMC9
Gene Name
ER membrane protein complex subunit 9
There are no transcripts associated with this gene.

Gene
FITM1
Gene Name
fat storage inducing transmembrane protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203402.2 498 Missense Mutation ATG,GTG M134V NP_981947.1
Gene
PSME1
Gene Name
proteasome activator subunit 1
There are no transcripts associated with this gene.

View Full Product Details