Product Details

SNP ID
rs9963473
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:35369591 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGAAGGATATTGGAAACATTGCAA[A/T]GCAGTTCTATGCCTAAAGAAGTCTT
Phenotype
MIM: 609600
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF396 PubMed Links

Gene Details

Gene
ZNF396
Gene Name
zinc finger protein 396
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322286.1 764 Missense Mutation CAT,CTT H211L NP_001309215.1
NM_001322290.1 764 Missense Mutation CAT,CTT H211L NP_001309219.1
NM_001322291.1 764 Intron NP_001309220.1
NM_145756.2 764 Missense Mutation CAT,CTT H211L NP_665699.1
XM_006722432.3 764 Missense Mutation CAT,CTT H211L XP_006722495.1
XM_017025687.1 764 Missense Mutation CAT,CTT H211L XP_016881176.1
XM_017025688.1 764 Missense Mutation CAT,CTT H211L XP_016881177.1
XM_017025689.1 764 Missense Mutation CAT,CTT H211L XP_016881178.1

View Full Product Details