Product Details

SNP ID
rs9842091
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:38266019 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGCCCCACCACTGTGCAGTGGC[C/T]TGGGTGAAGAACCACACTTTCAACC
Phenotype
MIM: 604047
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC22A13 PubMed Links

Gene Details

Gene
SLC22A13
Gene Name
solute carrier family 22 member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004256.3 114 Silent Mutation GCC,GCT A53A NP_004247.2
XM_017007519.1 114 UTR 5 XP_016863008.1

View Full Product Details