Product Details
- SNP ID
-
rs79339000
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:37993580 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTCCTAATAAAAGTCATGTTCTAT[A/G]ATGAAGTTGTACAGGTAGCCAGGTG
- Phenotype
-
MIM: 608592
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CTDSPL
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs542152054] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CTDSPL
- Gene Name
- CTD small phosphatase like
There are no transcripts associated with this gene.
- Gene
- VILL
- Gene Name
- villin like
View Full Product Details