Product Details
- SNP ID
-
hCV25960011
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:104092480 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTCAGCCGCCTGCCCTCTGAGGGG[G/T]CCCAGGGGCAGCCATAGCCCTTCCT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ASPG
PubMed Links
Gene Details
- Gene
- ASPG
- Gene Name
- asparaginase
- Gene
- LOC105370691
- Gene Name
- uncharacterized LOC105370691
There are no transcripts associated with this gene.
View Full Product Details