Product Details

SNP ID
rs3786948
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2933482 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACACTCTCCCAGGTCCACTGTATT[C/T]GTAGATTCACGCTCCAGCATGTGTT
Phenotype
MIM: 194551
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF77 PubMed Links

Gene Details

Gene
ZNF77
Gene Name
zinc finger protein 77
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021217.2 1801 UTR 3 NP_067040.1
XM_017027081.1 1801 UTR 3 XP_016882570.1
XM_017027082.1 1801 UTR 3 XP_016882571.1

View Full Product Details