Product Details

SNP ID
hCV25961449
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:127942572 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAAAGGGAGGGTATCATAAGCTC[G/T]GAAATATTTTTCTATGTCAACCCCA
Phenotype
MIM: 609697
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SAP130 PubMed Links

Gene Details

Gene
SAP130
Gene Name
Sin3A associated protein 130
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145928.1 Intron NP_001139400.1
NM_024545.3 Intron NP_078821.2

View Full Product Details