Product Details

SNP ID
rs8192620
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:132645140 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTTCAAGTAGCCAAACCAAATCAA[C/T]ACATCATTCAAAGTAGGTGGAATAA
Phenotype
MIM: 609333
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAAR1 PubMed Links

Gene Details

Gene
TAAR1
Gene Name
trace amine associated receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138327.2 887 Silent Mutation GTA,GTG V288V NP_612200.1

View Full Product Details