Product Details

SNP ID
rs201542780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50476541 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCCCTTCGCTGGTGGGAAGAAGC[C/G]GAGATGGCGGCAGCCAGCGCTGGGG
Phenotype
MIM: 614545
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EMC10 PubMed Links

Gene Details

Gene
EMC10
Gene Name
ER membrane protein complex subunit 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175063.5 206 UTR 5 NP_778233.4
NM_206538.3 206 UTR 5 NP_996261.1
XM_006723163.3 206 UTR 5 XP_006723226.1
XM_011526818.2 206 UTR 5 XP_011525120.1
Gene
FAM71E1
Gene Name
family with sequence similarity 71 member E1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308429.1 206 UTR 5 NP_001295358.1
NM_138411.2 206 UTR 5 NP_612420.1
XM_011526405.2 206 Intron XP_011524707.2
XM_011526406.2 206 Intron XP_011524708.2

View Full Product Details