Product Details

SNP ID
rs112214988
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:372345 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGAGACACAGGACATGTGGGGCA[C/T]GGCACAGCTTGTCTGTGTGAGCTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
B4GALNT4 PubMed Links

Gene Details

Gene
B4GALNT4
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178537.4 Intron NP_848632.2
XM_017017654.1 Intron XP_016873143.1

View Full Product Details