Product Details

SNP ID
rs151042142
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:85282126 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAATACTCAAAGTTTTTAGTTCA[C/T]TTGATATTTTACCTCAATTTTAAAA
Phenotype
MIM: 300603 MIM: 314990
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POF1B PubMed Links

Gene Details

Gene
POF1B
Gene Name
premature ovarian failure, 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307940.1 1987 UTR 3 NP_001294869.1
NM_024921.3 1987 Intron NP_079197.3
XM_005262203.3 1987 Intron XP_005262260.1
Gene
ZNF711
Gene Name
zinc finger protein 711
There are no transcripts associated with this gene.

View Full Product Details