Product Details

SNP ID
rs73505270
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4324576 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGAGACTGCAGTGGCATAGAGGG[A/G]ATGGCCAGTCCCCACTCAGCTGCCC
Phenotype
MIM: 608539
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLIS2 PubMed Links

Gene Details

Gene
GLIS2
Gene Name
GLIS family zinc finger 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318918.1 Intron NP_001305847.1
NM_032575.2 Intron NP_115964.2
XM_005255641.4 Intron XP_005255698.1
Gene
GLIS2-AS1
Gene Name
GLIS2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details