Product Details

SNP ID
rs71526500
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64826457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAATAGGGGACGGACAAGCCTCC[C/T]GGACCCGCTCACCTGCATTAGGGAG
Phenotype
MIM: 613991
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CDC42BPG PubMed Links

Gene Details

Gene
CDC42BPG
Gene Name
CDC42 binding protein kinase gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017525.2 Intron NP_059995.2
XM_011545155.2 Intron XP_011543457.1
XM_011545156.2 Intron XP_011543458.1
XM_011545157.2 Intron XP_011543459.1
XM_011545158.2 Intron XP_011543460.1
XM_011545159.2 Intron XP_011543461.1
XM_011545160.2 Intron XP_011543462.1
XM_011545161.2 Intron XP_011543463.1
XM_017017996.1 Intron XP_016873485.1
XM_017017997.1 Intron XP_016873486.1
XM_017017998.1 Intron XP_016873487.1
XM_017017999.1 Intron XP_016873488.1

View Full Product Details