Product Details

SNP ID
rs116337716
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39162707 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACCCTTCAGCCCTGGTGCCCTCC[C/T]CACCTCAGTGTTGGAGTAGTATGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARL5C PubMed Links

Gene Details

Gene
ARL5C
Gene Name
ADP ribosylation factor like GTPase 5C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143968.1 Intron NP_001137440.1

View Full Product Details