Product Details

SNP ID
rs34661454
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41807940 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATACACCAGGTTCTGCTGCATGAC[A/G]CTGTCCTTGGGGTCGAAGAGCATGT
Phenotype
MIM: 607063
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FKBP10 PubMed Links

Gene Details

Gene
FKBP10
Gene Name
FK506 binding protein 10
There are no transcripts associated with this gene.

Gene
P3H4
Gene Name
prolyl 3-hydroxylase family member 4 (non-enzymatic)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006455.2 1683 Intron NP_006446.1
XM_006721640.3 1683 Silent Mutation AGC,AGT S425S XP_006721703.1

View Full Product Details