Product Details

SNP ID
rs80027616
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:36991362 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGACCCCAGGGATGACTCGTACC[A/T]CTTCCAAGTCCACCCCGGCTCTGTC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TEX33 PubMed Links

Gene Details

Gene
TEX33
Gene Name
testis expressed 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163857.1 Intron NP_001157329.1
NM_178552.3 Intron NP_848647.1
XM_011530165.2 Intron XP_011528467.1
XM_011530166.1 Intron XP_011528468.1

View Full Product Details