Product Details

SNP ID
rs61747445
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141180215 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCGGCCTCGGTGGGTCGCTGCTC[A/G]GTGCCTGAGGGCCCCTTTCCAGGGC
Phenotype
MIM: 606345 MIM: 606333 MIM: 606334 MIM: 606335
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCDHB16 PubMed Links

Gene Details

Gene
PCDHB16
Gene Name
protocadherin beta 16
There are no transcripts associated with this gene.

Gene
PCDHB7
Gene Name
protocadherin beta 7
There are no transcripts associated with this gene.

Gene
PCDHB8
Gene Name
protocadherin beta 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019120.4 2426 Silent Mutation TCA,TCG S727S NP_061993.3
Gene
PCDHB9
Gene Name
protocadherin beta 9
There are no transcripts associated with this gene.

View Full Product Details