Product Details

SNP ID
rs11702061
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:37067386 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAATGCAACTGCCCAATATCTGCC[A/G]AAGAGAATATTAAAGTACATAATAG
Phenotype
MIM: 605938 MIM: 602259
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105372795 PubMed Links

Gene Details

Gene
LOC105372795
Gene Name
uncharacterized LOC105372795
There are no transcripts associated with this gene.

Gene
PIGP
Gene Name
phosphatidylinositol glycan anchor biosynthesis class P
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320480.1 Intron NP_001307409.1
NM_016430.3 Intron NP_057514.2
NM_153681.2 Intron NP_710148.1
NM_153682.2 Intron NP_710149.1
XM_005260990.4 Intron XP_005261047.1
XM_011529596.2 Intron XP_011527898.1
XM_017028364.1 Intron XP_016883853.1
XM_017028365.1 Intron XP_016883854.1
Gene
TTC3
Gene Name
tetratricopeptide repeat domain 3
There are no transcripts associated with this gene.

View Full Product Details