Product Details
- SNP ID
-
rs115224425
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:1525514 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTGGTCCCAGCCCTGGGTGGCCA[C/T]GAGATCGTACAGGAGGCAGGCGAGC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PLK5
PubMed Links
Gene Details
- Gene
- PLK5
- Gene Name
- polo like kinase 5
View Full Product Details