Product Details

SNP ID
rs61738060
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141234736 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATGTGGAAGACCTGGCTGCAAG[G/T]GGGGGCCAGAGTCATCTTTGACGAC
Phenotype
MIM: 606340
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PCDHB14 PubMed Links

Gene Details

Gene
PCDHB14
Gene Name
protocadherin beta 14
There are no transcripts associated with this gene.

Gene
PCDHB18P
Gene Name
protocadherin beta 18 pseudogene
There are no transcripts associated with this gene.

Gene
PCDHB19P
Gene Name
protocadherin beta 19 pseudogene
There are no transcripts associated with this gene.

View Full Product Details