Product Details

SNP ID
rs3767306
Assay Type
Functionally tested
NCBI dbSNP Submissions
47
Location
Chr.1:28209700 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GGGTAAGTGGCAATATGTGATGAGA[A/G]GGAGAGAACAACCCTTTCTCCCCAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJC8 PubMed Links

Gene Details

Gene
DNAJC8
Gene Name
DnaJ heat shock protein family (Hsp40) member C8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014280.2 Intron NP_055095.2

View Full Product Details