Product Details

SNP ID
rs75429031
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42314170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCTGGTCTCCTTCTAGTACTTG[A/G]GAAGCCAGGTACCTGGGGGGTAGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRR19 PubMed Links

Gene Details

Gene
PRR19
Gene Name
proline rich 19
There are no transcripts associated with this gene.

Gene
TMEM145
Gene Name
transmembrane protein 145
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173633.2 Intron NP_775904.2
XM_005258781.4 Intron XP_005258838.1
XM_011526791.2 Intron XP_011525093.1
XM_011526792.1 Intron XP_011525094.1

View Full Product Details