Product Details

SNP ID
rs61733852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63506423 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGCCCAGTGGGGGCAGGGCTCCT[G/A]GAACGAGGATCTACAGTTGGAGTTG
Phenotype
MIM: 606096
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
LGALS12 PubMed Links

Gene Details

Gene
LGALS12
Gene Name
galectin 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142535.1 372 Missense Mutation AGA,GGA R11G NP_001136007.1
NM_001142536.1 372 Missense Mutation AGA,GGA R11G NP_001136008.1
NM_001142537.1 372 Intron NP_001136009.1
NM_001142538.1 372 Intron NP_001136010.1
NM_033101.3 372 Missense Mutation AGA,GGA R11G NP_149092.2
XM_017018458.1 372 Missense Mutation AGA,GGA R11G XP_016873947.1

View Full Product Details