Product Details

SNP ID
rs61734934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:123906561 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGTTACTGTGGTGGGAAACCTC[A/C]GCATGATCTCAATTATTAGGCTGAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR8D4 PubMed Links

Gene Details

Gene
OR8D4
Gene Name
olfactory receptor family 8 subfamily D member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005197.1 130 Missense Mutation AGC,CGC S44R NP_001005197.1

View Full Product Details