Product Details

SNP ID
rs61753576
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:72012100 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGGGGGGAGGAGGACTTCTCTT[C/T]GCCTCCTACCTGATGGCCACGGGAG
Phenotype
MIM: 126064
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DHODH PubMed Links

Gene Details

Gene
DHODH
Gene Name
dihydroorotate dehydrogenase (quinone)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001361.4 72 Silent Mutation TTC,TTT F24F NP_001352.2
XM_005255827.3 72 UTR 5 XP_005255884.1
XM_005255829.3 72 Intron XP_005255886.1
XM_017022990.1 72 UTR 5 XP_016878479.1

View Full Product Details