Product Details

SNP ID
rs373555881
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:119127151 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAGAGAGCAGGACACAGGAAGGG[A/G]AGGAGCTCAAGGAAAGGTGGAAAAT
Phenotype
MIM: 607099
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2CD2L PubMed Links

Gene Details

Gene
C2CD2L
Gene Name
C2CD2 like
There are no transcripts associated with this gene.

Gene
HINFP
Gene Name
histone H4 transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243259.1 Intron NP_001230188.1
NM_015517.4 Intron NP_056332.2
NM_198971.2 Intron NP_945322.1
XM_011542744.2 Intron XP_011541046.1
XM_011542745.2 Intron XP_011541047.1
XM_011542746.1 Intron XP_011541048.1
XM_017017499.1 Intron XP_016872988.1
XM_017017500.1 Intron XP_016872989.1
XM_017017501.1 Intron XP_016872990.1
XM_017017502.1 Intron XP_016872991.1
XM_017017503.1 Intron XP_016872992.1
XM_017017504.1 Intron XP_016872993.1
XM_017017505.1 Intron XP_016872994.1
XM_017017506.1 Intron XP_016872995.1
XM_017017507.1 Intron XP_016872996.1
XM_017017508.1 Intron XP_016872997.1
XM_017017509.1 Intron XP_016872998.1

View Full Product Details