Product Details

SNP ID
rs41274953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:100302307 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAACACTTCCACTCTTCAATGTCT[A/G]GAGAGAAAAACAAGAGTAATCTGAG
Phenotype
MIM: 243305 MIM: 616717
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INVS PubMed Links

Gene Details

Gene
INVS
Gene Name
inversin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318381.1 Intron NP_001305310.1
NM_001318382.1 Intron NP_001305311.1
NM_014425.4 Intron NP_055240.2
Gene
TEX10
Gene Name
testis expressed 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161584.1 Intron NP_001155056.1
NM_017746.3 Intron NP_060216.2
XM_011518798.1 Intron XP_011517100.1
XM_017014848.1 Intron XP_016870337.1

View Full Product Details