Product Details
- SNP ID
-
hCV25984688
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:125074062 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGCTAATTTTGGGGAATACTGAAT[C/T]GTATCTCCTTTTTACAACCTCATGC
- Phenotype
-
MIM: 614364
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AACS
PubMed Links
Gene Details
- Gene
- AACS
- Gene Name
- acetoacetyl-CoA synthetase
There are no transcripts associated with this gene.
View Full Product Details