Product Details

SNP ID
rs61752498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:23289136 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCTGGAAGAGGAGGACGGTCAT[C/T]TGTCACCAGGACATCAGGTGTGTAG
Phenotype
MIM: 606663
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOXL2 PubMed Links

Gene Details

Gene
LOXL2
Gene Name
lysyl oxidase like 2
There are no transcripts associated with this gene.

Gene
R3HCC1
Gene Name
R3H domain and coiled-coil containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136108.2 315 Silent Mutation ATC,ATT I77I NP_001129580.2
NM_001301650.1 315 Silent Mutation ATC,ATT I77I NP_001288579.1

View Full Product Details