Product Details

SNP ID
rs28448343
Assay Type
Functionally tested
NCBI dbSNP Submissions
51
Location
Chr.1:248349196 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCGTGAACTCTCGAATCTGCATCC[A/G]GATGACACTGGCCTCCCTACTCAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR14C36 PubMed Links

Gene Details

Gene
OR14C36
Gene Name
olfactory receptor family 14 subfamily C member 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001918.1 422 Missense Mutation CAG,CGG Q141R NP_001001918.1

View Full Product Details