Product Details

SNP ID
rs11547584
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:9365020 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATTTCTTTTTCTTTTAAAGGACA[A/G]TACTGCTTTTAAAGAGACAGTGTTA
Phenotype
MIM: 606525
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SRGAP3 PubMed Links

Gene Details

Gene
SRGAP3
Gene Name
SLIT-ROBO Rho GTPase activating protein 3
There are no transcripts associated with this gene.

Gene
THUMPD3
Gene Name
THUMP domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114092.1 662 UTR 5 NP_001107564.1
NM_015453.2 662 UTR 5 NP_056268.2
XM_005265021.4 662 UTR 5 XP_005265078.1
XM_005265022.4 662 UTR 5 XP_005265079.1
XM_005265023.3 662 UTR 5 XP_005265080.1
XM_005265024.4 662 UTR 5 XP_005265081.1
XM_006713086.3 662 UTR 5 XP_006713149.1
XM_011533570.2 662 UTR 5 XP_011531872.1
XM_011533571.2 662 UTR 5 XP_011531873.1
XM_017006113.1 662 UTR 5 XP_016861602.1
XM_017006114.1 662 UTR 5 XP_016861603.1
XM_017006115.1 662 UTR 5 XP_016861604.1
XM_017006116.1 662 UTR 5 XP_016861605.1
XM_017006117.1 662 UTR 5 XP_016861606.1
XM_017006118.1 662 UTR 5 XP_016861607.1
XM_017006119.1 662 UTR 5 XP_016861608.1

View Full Product Details