Product Details

SNP ID
rs61753924
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35142731 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGTTCCCCCCAGGTCTGTCCACC[C/T]GCAGGCGGTAGAAACACCTGGAGCG
Phenotype
MIM: 602359 MIM: 606684 MIM: 608303
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FXYD1 PubMed Links

Gene Details

Gene
FXYD1
Gene Name
FXYD domain containing ion transport regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278717.1 357 Missense Mutation CGC,TGC R90C NP_001265646.1
NM_001278718.1 357 Missense Mutation CGC,TGC R90C NP_001265647.1
NM_005031.4 357 Missense Mutation CGC,TGC R90C NP_005022.2
NM_021902.3 357 Missense Mutation CGC,TGC R90C NP_068702.1
XM_017026874.1 357 Missense Mutation CGC,TGC R90C XP_016882363.1
XM_017026875.1 357 Missense Mutation CGC,TGC R108C XP_016882364.1
XM_017026876.1 357 Missense Mutation CGC,TGC R108C XP_016882365.1
Gene
FXYD7
Gene Name
FXYD domain containing ion transport regulator 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022006.1 357 Intron NP_071289.1
Gene
LGI4
Gene Name
leucine rich repeat LGI family member 4
There are no transcripts associated with this gene.

View Full Product Details