Product Details

SNP ID
rs17221305
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:7849213 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTCAGCTGTCTTTCTGCAACTCC[A/G]ACGTCATCCACCACTTCTACTGTGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC283299 PubMed Links

Gene Details

Gene
LOC283299
Gene Name
uncharacterized LOC283299
There are no transcripts associated with this gene.

Gene
OR5E1P
Gene Name
olfactory receptor family 5 subfamily E member 1 pseudogene
There are no transcripts associated with this gene.

View Full Product Details