Product Details

SNP ID
rs9467165
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:10671733 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGTAAAGGTTAAAATTCTTAAAT[G/T]GCATATTTCCTTTCAATAAGACTTG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C6orf52 PubMed Links

Gene Details

Gene
C6orf52
Gene Name
chromosome 6 open reading frame 52
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145020.1 Intron NP_001138492.1
XM_006715088.3 Intron XP_006715151.1
XM_011514572.2 Intron XP_011512874.1
XM_011514574.2 Intron XP_011512876.1
XM_011514575.2 Intron XP_011512877.1

View Full Product Details