Product Details
- SNP ID
-
rs192396803
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:100189814 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACTGGCCCTGCACAGTGGGCTGCC[C/T]CATTCGTGTCCTAGACTCCATGCCC
- Phenotype
-
MIM: 611030
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM30C
PubMed Links
Gene Details
- Gene
- TMEM30C
- Gene Name
- transmembrane protein 30C
There are no transcripts associated with this gene.
View Full Product Details