Product Details

SNP ID
rs73560777
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46652904 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGAGATGGCGTGGGGAGAGACAC[A/G]GGGGGCCTCAGAAATAGGGATACGG
Phenotype
MIM: 611112
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DACT3 PubMed Links

Gene Details

Gene
DACT3
Gene Name
dishevelled binding antagonist of beta catenin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301046.1 Intron NP_001287975.1
NM_145056.2 Intron NP_659493.2
XM_011526499.2 Intron XP_011524801.1
XM_011526500.2 Intron XP_011524802.1
XM_017026334.1 Intron XP_016881823.1
Gene
DACT3-AS1
Gene Name
DACT3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details