Product Details

SNP ID
rs60744039
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:70809138 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCAGCATCCCTTCCTGGTGCTGT[A/G]GCTCCTAGGGAGCTGACATGGGATG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CLEC4F PubMed Links

Gene Details

Gene
CLEC4F
Gene Name
C-type lectin domain family 4 member F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258027.1 1980 UTR 3 NP_001244956.1
NM_001321308.1 1980 Missense Mutation CAC,TAC H555Y NP_001308237.1
NM_173535.2 1980 UTR 3 NP_775806.2
XM_011532635.2 1980 UTR 3 XP_011530937.1
XM_011532637.2 1980 UTR 3 XP_011530939.1
XM_011532638.2 1980 Silent Mutation GCC,GCT A667A XP_011530940.1
XM_011532639.2 1980 Missense Mutation CAC,TAC H665Y XP_011530941.1
XM_011532640.2 1980 UTR 3 XP_011530942.1
XM_011532641.2 1980 Intron XP_011530943.1
XM_011532642.2 1980 UTR 3 XP_011530944.1
XM_011532643.1 1980 UTR 3 XP_011530945.1
XM_017003519.1 1980 UTR 3 XP_016859008.1

View Full Product Details