Product Details
- SNP ID
-
hCV25991064
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:54634791 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGAAAGGCCATACTCACCAGCATA[A/G]ACCAGGGCCCCTGCTACAGCTGCCA
- Phenotype
-
MIM: 606634
MIM: 607360
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DCD
PubMed Links
Gene Details
- Gene
- DCD
- Gene Name
- dermcidin
There are no transcripts associated with this gene.
- Gene
- LACRT
- Gene Name
- lacritin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_033277.1 |
89 |
Silent Mutation |
GTC,GTT |
V17V |
NP_150593.1 |
View Full Product Details