Product Details

SNP ID
rs117782866
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67989266 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGGACGTGATGGCTATGGTGACC[A/G]TGAAGGCCCCACTGCTGTACAGCTA
Phenotype
MIM: 609925 MIM: 609926
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPEP2 PubMed Links

Gene Details

Gene
DPEP2
Gene Name
dipeptidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324159.1 Intron NP_001311088.1
NM_022355.3 Intron NP_071750.1
XM_005256090.4 Intron XP_005256147.2
XM_011523266.1 Intron XP_011521568.1
XM_011523268.2 Intron XP_011521570.1
XM_011523271.2 Intron XP_011521573.1
XM_011523272.2 Intron XP_011521574.1
XM_011523273.2 Intron XP_011521575.1
XM_011523274.2 Intron XP_011521576.1
XM_017023541.1 Intron XP_016879030.1
XM_017023542.1 Intron XP_016879031.1
XM_017023543.1 Intron XP_016879032.1
XM_017023544.1 Intron XP_016879033.1
XM_017023545.1 Intron XP_016879034.1
XM_017023546.1 Intron XP_016879035.1
XM_017023547.1 Intron XP_016879036.1
Gene
DPEP3
Gene Name
dipeptidase 3
There are no transcripts associated with this gene.

View Full Product Details