Product Details

SNP ID
rs8049883
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56639498 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGACGTGATGGATTCCCAAGTTTC[A/G]TTCTTAAAATAGACAAACTGAGGCC
Phenotype
MIM: 156350 MIM: 156356 MIM: 156357
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MT1A PubMed Links

Gene Details

Gene
MT1A
Gene Name
metallothionein 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005946.2 Intron NP_005937.2
Gene
MT1DP
Gene Name
metallothionein 1D, pseudogene
There are no transcripts associated with this gene.

Gene
MT1JP
Gene Name
metallothionein 1J, pseudogene
There are no transcripts associated with this gene.

Gene
MT1M
Gene Name
metallothionein 1M
There are no transcripts associated with this gene.

View Full Product Details