Product Details
- SNP ID
-
rs1133090
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:67987565 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACAACTGCAAGGACTGGGGCCATGT[C/G]GGGGGAGGACTCTGAGACTGACCAC
- Phenotype
-
MIM: 609925
MIM: 609926
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DPEP2
PubMed Links
Gene Details
- Gene
- DPEP2
- Gene Name
- dipeptidase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001324159.1 |
729 |
Missense Mutation |
CAC,GAC |
H310D |
NP_001311088.1 |
NM_022355.3 |
729 |
Missense Mutation |
CAC,GAC |
H468D |
NP_071750.1 |
XM_005256090.4 |
729 |
UTR 3 |
|
|
XP_005256147.2 |
XM_011523266.1 |
729 |
Missense Mutation |
CAC,GAC |
H481D |
XP_011521568.1 |
XM_011523268.2 |
729 |
Missense Mutation |
CAC,GAC |
H473D |
XP_011521570.1 |
XM_011523271.2 |
729 |
UTR 3 |
|
|
XP_011521573.1 |
XM_011523272.2 |
729 |
Missense Mutation |
CAC,GAC |
H323D |
XP_011521574.1 |
XM_011523273.2 |
729 |
Missense Mutation |
CAC,GAC |
H323D |
XP_011521575.1 |
XM_011523274.2 |
729 |
Missense Mutation |
CAC,GAC |
H273D |
XP_011521576.1 |
XM_017023541.1 |
729 |
Missense Mutation |
CAC,GAC |
H415D |
XP_016879030.1 |
XM_017023542.1 |
729 |
Missense Mutation |
CAC,GAC |
H391D |
XP_016879031.1 |
XM_017023543.1 |
729 |
Missense Mutation |
CAC,GAC |
H365D |
XP_016879032.1 |
XM_017023544.1 |
729 |
Missense Mutation |
CAC,GAC |
H358D |
XP_016879033.1 |
XM_017023545.1 |
729 |
Missense Mutation |
CAC,GAC |
H323D |
XP_016879034.1 |
XM_017023546.1 |
729 |
Missense Mutation |
CAC,GAC |
H232D |
XP_016879035.1 |
XM_017023547.1 |
729 |
Missense Mutation |
CAC,GAC |
H232D |
XP_016879036.1 |
- Gene
- DPEP3
- Gene Name
- dipeptidase 3
There are no transcripts associated with this gene.
View Full Product Details